Ir[tBu(tBu)ppy]2(4,4'-dCF3bpy)PF6 97%

Supplier: Ambeed
Warning

A1495457-100MG A1495457-250MG A1495457-50MG
76666-418EA 236.59 USD
76666-418 76666-420 76666-422
Ir[tBu(tBu)ppy]2(4,4'-dCF3bpy)PF6 97%
Ir[tBu(tBu)ppy]2(4,4'-dCF3bpy)PF6

Precaución: for Research Use Only

Formula: C₅₀H₅₄F₁₂IrN₄P
MW: 1162.16 g/mol
CAS Number: 2757085-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR