Ir[FCF3(CF3)ppy]2(4,4'-dCF3bpy)PF6 97%

Supplier: Ambeed
Warning

A1495455-100MG A1495455-250MG A1495455-50MG A1495455-1G
76666-406EA 244 USD
76666-406 76666-408 76666-410 77875-626
Ir[FCF3(CF3)ppy]2(4,4'-dCF3bpy)PF6 97%
Ir[FCF3(CF3)ppy]2(4,4'-dCF3bpy)PF6

Precaución: for Research Use Only

Formula: C₃₈H₁₆F₂₀IrN₄
CAS Number: 2757085-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR