[5,5'-Bis-trifluoromethyl]bis[(5-fluoro-2-(5-methyl-2-pyridinyl)phenyl]iridium(III) hexafluorophosphate 97%

Supplier: Ambeed
Warning

A1495453-100MG A1495453-1G A1495453-250MG A1495453-50MG
76666-392EA 247.19 USD
76666-392 76666-394 76666-396 76666-398
[5,5'-Bis-trifluoromethyl]bis[(5-fluoro-2-(5-methyl-2-pyridinyl)phenyl]iridium(III) hexafluorophosphate 97%
[5,5'-Bis-trifluoromethyl]bis[(5-fluoro-2-(5-methyl-2-pyridinyl)phenyl]iridium(III) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₆H₂₄F₁₄IrN₄P
MW: 1001.77 g/mol
CAS Number: 1191467-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR