[4,4'-Bis-trifluoromethyl]bis[(5-fluoro-2-(5-methyl-2-pyridinyl)phenyl]iridium(III) hexafluorophosphate 97%

Supplier: Ambeed
Warning

A1495452-100MG A1495452-1G A1495452-250MG A1495452-50MG
76666-384EA 221 USD
76666-384 76666-386 76666-388 76666-390
[4,4'-Bis-trifluoromethyl]bis[(5-fluoro-2-(5-methyl-2-pyridinyl)phenyl]iridium(III) hexafluorophosphate 97%
[4,4'-Bis-trifluoromethyl]bis[(5-fluoro-2-(5-methyl-2-pyridinyl)phenyl]iridium(III) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₆H₂₄F₁₄IrN₄P
CAS Number: 2773544-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR