[5,5'-Bis-trifluoromethyl]bis[(4-fluorophenyl)pyridine]iridium(III) hexafluorophosphate 97%

Supplier: Ambeed
Warning

A1495451-100MG A1495451-1G A1495451-250MG A1495451-50MG
76666-376EA 103 USD
76666-376 76666-378 76666-380 76666-382
[5,5'-Bis-trifluoromethyl]bis[(4-fluorophenyl)pyridine]iridium(III) hexafluorophosphate 97%
[5,5'-Bis-trifluoromethyl]bis[(4-fluorophenyl)pyridine]iridium(III) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₄H₂₀F₈IrN₄
MW: 828.7536 g/mol
CAS Number: 2757085-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR