[5,5'-Bis-trifluoromethyl]bis[2-(2,4-difluorophenyl)pyridine]iridium(III) hexafluorophosphate 97%

Supplier: Ambeed
Warning

A1495449-100MG A1495449-1G A1495449-250MG A1495449-50MG
76666-362EA 95 USD
76666-362 76666-364 76666-366 76666-368
[5,5'-Bis-trifluoromethyl]bis[2-(2,4-difluorophenyl)pyridine]iridium(III) hexafluorophosphate 97%
[5,5'-Bis-trifluoromethyl]bis[2-(2,4-difluorophenyl)pyridine]iridium(III) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₄H₁₈F₁₆IrN₄P
CAS Number: 2757084-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR