[4,4'-Bis-trifluoromethyl]bis[2-(2,4-difluorophenyl)pyridine]iridium(III) hexafluorophosphate 97%

Supplier: Ambeed
Warning

A1495448-100MG A1495448-1G A1495448-250MG A1495448-50MG
76666-354EA 245.07 USD
76666-354 76666-356 76666-358 76666-360
[4,4'-Bis-trifluoromethyl]bis[2-(2,4-difluorophenyl)pyridine]iridium(III) hexafluorophosphate 97%
[4,4'-Bis-trifluoromethyl]bis[2-(2,4-difluorophenyl)pyridine]iridium(III) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₄H₁₈F₁₀IrN₄
MW: 864.7346 g/mol
CAS Number: 2757084-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR