Methanesulfonato(2-dicyclohexylphosphino-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) 98+%

Supplier: Ambeed
Warning

Synonyms: Brettphos Pd G4

A1495336-100MG A1495336-1G A1495336-250MG A1495336-5G A1495336-25G
76666-340EA 8 USD
76666-340 76666-342 76666-344 76666-346 77597-698
Methanesulfonato(2-dicyclohexylphosphino-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) 98+%
Methanesulfonato(2-dicyclohexylphosphino-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₄₉H₆₈NO₅PPdS
MDL Number: MFCD29917502
CAS Number: 1599466-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR