1-(11,12-Didehydrodibenz[b,f]azocin-5(6H)-yl)ethanone 97%

Supplier: Ambeed
Warning

A1495298-1G A1495298-250MG
76666-332EA 247.19 USD
76666-332 76666-334
1-(11,12-Didehydrodibenz[b,f]azocin-5(6H)-yl)ethanone 97%
1-(11,12-Didehydrodibenz[b,f]azocin-5(6H)-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₇H₁₃NO
MW: 247.29 g/mol
CAS Number: 1421353-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR