Methyl 2-amino-5-(dimethylphosphoryl)benzoate 95%
Supplier: Ambeed
|
Warning
|
A1495228-100MG
A1495228-250MG
76666-324EA
111.4
USD
76666-324
76666-326
Methyl 2-amino-5-(dimethylphosphoryl)benzoate 95%
Methyl 2-amino-5-(dimethylphosphoryl)benzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₄NO₃P MW: 227.1968 g/mol |
CAS Number:
2503203-14-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...