Methyl 2-amino-4-(dimethylphosphoryl)benzoate 95%
Supplier: Ambeed
|
Warning
|
A1495227-100MG
A1495227-1G
A1495227-250MG
76666-318EA
20.16
USD
76666-318
76666-320
76666-322
Methyl 2-amino-4-(dimethylphosphoryl)benzoate 95%
Methyl 2-amino-4-(dimethylphosphoryl)benzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₄NO₃P MW: 227.1968 g/mol |
CAS Number:
2551116-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...