(4-Amino-3-fluorophenyl)dimethylphosphine oxide 95%

Supplier: Ambeed
Warning

A1495226-100MG A1495226-250MG A1495226-1G
76666-314EA 40 USD
76666-314 76666-316 77875-614
(4-Amino-3-fluorophenyl)dimethylphosphine oxide 95%
(4-Amino-3-fluorophenyl)dimethylphosphine oxide

Precaución: for Research Use Only

Formula: C₈H₁₁FNOP
CAS Number: 1197956-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR