4-(Dimethylphosphoryl)benzonitrile 95%

Supplier: Ambeed
Warning

A1495220-100MG A1495220-250MG
76666-288EA 20.16 USD
76666-288 76666-290
4-(Dimethylphosphoryl)benzonitrile 95%
4-(Dimethylphosphoryl)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₁₀NOP
MW: 179.1556 g/mol
MDL Number: MFCD33549332
CAS Number: 61738-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR