1-(4-(Dimethylphosphoryl)phenyl)ethan-1-one 95%

Supplier: Ambeed
Warning

A1495217-100MG A1495217-1G A1495217-250MG
76666-280EA 27.59 USD
76666-280 76666-282 76666-284
1-(4-(Dimethylphosphoryl)phenyl)ethan-1-one 95%
1-(4-(Dimethylphosphoryl)phenyl)ethan-1-one

Precaución: for Research Use Only

Formula: C₁₀H₁₃O₂P
MW: 196.18 g/mol
MDL Number: MFCD32875222
CAS Number: 2551117-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR