1-(3-(Dimethylphosphoryl)phenyl)ethan-1-one 95%

Supplier: Ambeed
Warning

A1495216-100MG A1495216-1G A1495216-250MG
76666-274EA 20.16 USD
76666-274 76666-276 76666-278
1-(3-(Dimethylphosphoryl)phenyl)ethan-1-one 95%
1-(3-(Dimethylphosphoryl)phenyl)ethan-1-one

Precaución: for Research Use Only

Formula: C₁₀H₁₃O₂P
MW: 196.1828 g/mol
CAS Number: 2551118-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR