(5-Bromopyrimidin-2-yl)dimethylphosphine oxide 95%

Supplier: Ambeed
Warning

A1495215-100MG A1495215-50MG
76666-270EA 269 USD
76666-270 76666-272
(5-Bromopyrimidin-2-yl)dimethylphosphine oxide 95%
(5-Bromopyrimidin-2-yl)dimethylphosphine oxide

Precaución: for Research Use Only

Formula: C₆H₈BrN₂OP
MDL Number: MFCD33032421
CAS Number: 2613382-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR