(2-Aminopyrimidin-5-yl)dimethylphosphine oxide 95%

Supplier: Ambeed
Warning

A1495212-100MG A1495212-1G A1495212-250MG A1495212-50MG
76666-254EA 57 USD
76666-254 76666-256 76666-258 76666-260
(2-Aminopyrimidin-5-yl)dimethylphosphine oxide 95%
(2-Aminopyrimidin-5-yl)dimethylphosphine oxide

Precaución: for Research Use Only

Formula: C₆H₁₀N₃OP
MDL Number: MFCD32874549
CAS Number: 2362008-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR