(2-Aminopyridin-3-yl)dimethylphosphine oxide 97%

Supplier: Ambeed
Danger

A1495196-250MG A1495196-100MG A1495196-1G
76666-248EA 216 USD
76666-248 77875-598 77875-599
(2-Aminopyridin-3-yl)dimethylphosphine oxide 97%
(2-Aminopyridin-3-yl)dimethylphosphine oxide

Precaución: for Research Use Only

Formula: C₇H₁₁N₂OP
MDL Number: MFCD32268255
CAS Number: 2229830-69-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR