4-(4-(1,2,2-Triphenylvinyl)phenyl)pyridine 98%

Supplier: Ambeed
Warning

A1494412-100MG A1494412-1G A1494412-250MG
76666-236EA 28 USD
76666-236 76666-238 76666-240
4-(4-(1,2,2-Triphenylvinyl)phenyl)pyridine 98%
4-(4-(1,2,2-Triphenylvinyl)phenyl)pyridine

Precaución: for Research Use Only

Formula: C₃₁H₂₃N
CAS Number: 1915012-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR