1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl acetate 93% + (by GC)

Supplier: Ambeed
Warning

A1494159-100G A1494159-500G
76666-224EA 9 USD
76666-224 76666-226 76666-228
1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl acetate 93% + (by GC)
1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl acetate

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₂
MDL Number: MFCD00135943
CAS Number: 92618-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR