Geranyl chloride 95%

Supplier: Ambeed
Warning

Synonyms: (E)-1-Chloro-3,7-dimethylocta-2,6-diene

A1494121-1G A1494121-250MG
76666-220EA 27.59 USD
76666-220 76666-222
Geranyl chloride 95%
Geranyl chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₇Cl
MW: 172.6950 g/mol
MDL Number: MFCD00000983
CAS Number: 5389-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR