(S)-4-Benzyl-2-((R)-1-(diphenylphosphanyl)piperidin-2-yl)-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1494062-100MG A1494062-250MG
76666-198EA 235.52 USD
76666-198 76666-200
(S)-4-Benzyl-2-((R)-1-(diphenylphosphanyl)piperidin-2-yl)-4,5-dihydrooxazole 97%
(S)-4-Benzyl-2-((R)-1-(diphenylphosphanyl)piperidin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₇H₂₉N₂OP
MW: 428.50 g/mol
CAS Number: 2757084-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR