(R)-2-(Benzo[b]thiophen-2-yl)-4-phenyl-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1493996-100MG A1493996-1G A1493996-250MG
76666-176EA 47 USD
76666-176 76666-178 76666-180
(R)-2-(Benzo[b]thiophen-2-yl)-4-phenyl-4,5-dihydrooxazole ≥97%, ee 99%
(R)-2-(Benzo[b]thiophen-2-yl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₇H₁₃NOS
CAS Number: 541549-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR