(S)-2-(Benzo[b]thiophen-2-yl)-4-benzyl-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1493991-100MG A1493991-1G A1493991-250MG A1493991-50MG
76666-162EA 69 USD
76666-162 76666-164 76666-166 76666-168
(S)-2-(Benzo[b]thiophen-2-yl)-4-benzyl-4,5-dihydrooxazole 97%
(S)-2-(Benzo[b]thiophen-2-yl)-4-benzyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₈H₁₅NOS
CAS Number: 2170033-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR