(S)-2-(Benzo[b]thiophen-2-yl)-4-phenyl-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1493990-100MG A1493990-1G A1493990-250MG
76666-156EA 74 USD
76666-156 76666-158 76666-160
(S)-2-(Benzo[b]thiophen-2-yl)-4-phenyl-4,5-dihydrooxazole ≥97%, ee 99%
(S)-2-(Benzo[b]thiophen-2-yl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₇H₁₃NOS
CAS Number: 2757083-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR