(S)-2-(Benzo[b]thiophen-2-yl)-4-(tert-butyl)-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1493989-100MG A1493989-250MG A1493989-1G
76666-152EA 124.13 USD
76666-152 76666-154 77624-542
(S)-2-(Benzo[b]thiophen-2-yl)-4-(tert-butyl)-4,5-dihydrooxazole 97%
(S)-2-(Benzo[b]thiophen-2-yl)-4-(tert-butyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₅H₁₇NOS
MW: 259.3666 g/mol
CAS Number: 541549-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR