(1R,2R)-1,2-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)cyclohexane 97%

Supplier: Ambeed
Warning

A1493977-100MG A1493977-1G A1493977-250MG
76666-124EA 118.83 USD
76666-124 76666-126 76666-128
(1R,2R)-1,2-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)cyclohexane 97%
(1R,2R)-1,2-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)cyclohexane

Precaución: for Research Use Only

Formula: C₂₀H₃₄N₂O₂
MW: 334.4962 g/mol
CAS Number: 2757083-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR