(4R,5S)-2-(5-Bromopyridin-2-yl)-4,5-diphenyl-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1493917-100MG A1493917-1G A1493917-250MG
76666-102EA 85 USD
76666-102 76666-104 76666-106
(4R,5S)-2-(5-Bromopyridin-2-yl)-4,5-diphenyl-4,5-dihydrooxazole 97%
(4R,5S)-2-(5-Bromopyridin-2-yl)-4,5-diphenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₀H₁₅BrN₂O
CAS Number: 2757083-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR