(3aR,8aS)-2-(5-Bromopyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A1493916-100MG
A1493916-1G
A1493916-250MG
A1493916-50MG
76666-094EA
43
USD
76666-094
76666-096
76666-098
76666-100
(3aR,8aS)-2-(5-Bromopyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole ≥97%, ee 99%
(3aR,8aS)-2-(5-Bromopyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole
Precaución: for Research Use Only
Formula:
C₁₅H₁₁BrN₂O |
CAS Number:
2757083-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...