(3aS,8aR)-2-(5-Bromopyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A1493910-100MG
A1493910-1G
A1493910-250MG
76666-070EA
65
USD
76666-070
76666-072
76666-074
(3aS,8aR)-2-(5-Bromopyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole ≥97%, ee 99%
(3aS,8aR)-2-(5-Bromopyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole
Precaución: for Research Use Only
Formula:
C₁₅H₁₁BrN₂O |
CAS Number:
2757083-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...