(S)-2-(5-Bromopyridin-2-yl)-4-phenyl-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1493908-100MG A1493908-250MG A1493908-1G
76666-062EA 53 USD
76666-062 76666-064 77875-516
(S)-2-(5-Bromopyridin-2-yl)-4-phenyl-4,5-dihydrooxazole ≥97%, ee 99%
(S)-2-(5-Bromopyridin-2-yl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrN₂O
CAS Number: 2757083-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR