(S)-2-(5-Bromopyridin-2-yl)-4-(tert-butyl)-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1493907-100MG A1493907-250MG A1493907-1G
76666-058EA 53 USD
76666-058 76666-060 77875-515
(S)-2-(5-Bromopyridin-2-yl)-4-(tert-butyl)-4,5-dihydrooxazole ≥97%, ee 99%
(S)-2-(5-Bromopyridin-2-yl)-4-(tert-butyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrN₂O
CAS Number: 2757083-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR