(S)-2-(5-Bromopyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1493906-100MG A1493906-250MG A1493906-1G
76666-054EA 142 USD
76666-054 76666-056 77875-514
(S)-2-(5-Bromopyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole ≥97%, ee 99%
(S)-2-(5-Bromopyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrN₂O
CAS Number: 2757083-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR