(R)-2-(5-Chloropyridin-2-yl)-4-phenyl-4,5-dihydrooxazole 97%
Supplier: Ambeed
|
Warning
|
A1493902-100MG
A1493902-250MG
76666-046EA
150.65
USD
76666-046
76666-048
(R)-2-(5-Chloropyridin-2-yl)-4-phenyl-4,5-dihydrooxazole 97%
(R)-2-(5-Chloropyridin-2-yl)-4-phenyl-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₁₄H₁₁ClN₂O MW: 258.7029 g/mol |
CAS Number:
2828433-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...