(S)-4-(tert-Butyl)-2-(5-chloropyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1493895-100MG A1493895-250MG
76666-022EA 150.65 USD
76666-022 76666-024
(S)-4-(tert-Butyl)-2-(5-chloropyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%
(S)-4-(tert-Butyl)-2-(5-chloropyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClN₂O
MW: 238.7133 g/mol
CAS Number: 2757083-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR