(4R,5S)-2-(5-Fluoropyridin-2-yl)-4,5-diphenyl-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1493893-100MG A1493893-1G A1493893-250MG
76666-010EA 150.65 USD
76666-010 76666-012 76666-014
(4R,5S)-2-(5-Fluoropyridin-2-yl)-4,5-diphenyl-4,5-dihydrooxazole 97%
(4R,5S)-2-(5-Fluoropyridin-2-yl)-4,5-diphenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₀H₁₅FN₂O
MW: 318.34 g/mol
CAS Number: 2757083-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR