(R)-4-Benzyl-2-(5-fluoropyridin-2-yl)-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1493891-250MG A1493891-100MG
76666-008EA 255.68 USD
76666-008 77624-532
(R)-4-Benzyl-2-(5-fluoropyridin-2-yl)-4,5-dihydrooxazole 97%
(R)-4-Benzyl-2-(5-fluoropyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₅H₁₃FN₂O
MW: 256.2749 g/mol
CAS Number: 2757083-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR