(R)-4-(tert-Butyl)-2-(5-fluoropyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1493889-250MG A1493889-1G
76666-006EA 241 USD
76666-006 77875-507
(R)-4-(tert-Butyl)-2-(5-fluoropyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%
(R)-4-(tert-Butyl)-2-(5-fluoropyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₂H₁₅FN₂O
CAS Number: 2757083-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR