(R)-2-(5-Fluoropyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A1493888-100MG
A1493888-1G
A1493888-250MG
76666-000EA
150.65
USD
76666-000
76666-002
76666-004
(R)-2-(5-Fluoropyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole ≥97%, ee 99%
(R)-2-(5-Fluoropyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₁₁H₁₃FN₂O MW: 208.2321 g/mol |
CAS Number:
2828433-61-8 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...