(S)-4-Benzyl-2-(5-fluoropyridin-2-yl)-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1493885-100MG A1493885-1G A1493885-250MG
76665-994EA 150.65 USD
76665-994 76665-996 76665-998
(S)-4-Benzyl-2-(5-fluoropyridin-2-yl)-4,5-dihydrooxazole 97%
(S)-4-Benzyl-2-(5-fluoropyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₅H₁₃FN₂O
MW: 256.2749 g/mol
CAS Number: 2757083-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR