(S)-2-(5-Fluoropyridin-2-yl)-4-phenyl-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1493884-100MG A1493884-250MG A1493884-50MG
76665-988EA 150.65 USD
76665-988 76665-990 76665-992
(S)-2-(5-Fluoropyridin-2-yl)-4-phenyl-4,5-dihydrooxazole 97%
(S)-2-(5-Fluoropyridin-2-yl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₄H₁₁FN₂O
MW: 242.2484 g/mol
MDL Number: MFCD34596715
CAS Number: 2757083-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR