(S)-4-(tert-Butyl)-2-(5-fluoropyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1493883-100MG A1493883-250MG A1493883-50MG
76665-982EA 141.1 USD
76665-982 76665-984 76665-986
(S)-4-(tert-Butyl)-2-(5-fluoropyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%
(S)-4-(tert-Butyl)-2-(5-fluoropyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₂H₁₅FN₂O
MW: 222.2587 g/mol
CAS Number: 2068819-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR