(S)-2-(5-Fluoropyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1493882-100MG A1493882-250MG
76665-978EA 150.65 USD
76665-978 76665-980
(S)-2-(5-Fluoropyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole ≥97%, ee 99%
(S)-2-(5-Fluoropyridin-2-yl)-4-isopropyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₁H₁₃FN₂O
MW: 208.23 g/mol
CAS Number: 2757083-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR