(E)-3-(3,4-Dichlorophenyl)-N-(2-(4-(methylsulfonyl)piperazin-1-yl)-2-oxoethyl)acrylamide 95%

Supplier: Ambeed
Warning

A1493794-100MG A1493794-25MG A1493794-50MG A1493794-250MG
76665-972EA 326 USD
76665-972 76665-974 76665-976 77875-503
(E)-3-(3,4-Dichlorophenyl)-N-(2-(4-(methylsulfonyl)piperazin-1-yl)-2-oxoethyl)acrylamide 95%
(E)-3-(3,4-Dichlorophenyl)-N-(2-(4-(methylsulfonyl)piperazin-1-yl)-2-oxoethyl)acrylamide

Precaución: for Research Use Only

Formula: C₁₆H₁₉Cl₂N₃O₄S
MDL Number: MFCD32693929
CAS Number: 2505001-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR