trans,trans-4'-Ethyl-4-(3,4,5-trifluorophenyl)bicyclohexyl 98%

Supplier: Ambeed
Warning

Synonyms: trans(trans)-4-Ethyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane)

A1493576-100G A1493576-25G A1493576-5G
76665-966EA 137.92 USD
76665-966 76665-968 76665-970
trans,trans-4'-Ethyl-4-(3,4,5-trifluorophenyl)bicyclohexyl 98%
trans,trans-4'-Ethyl-4-(3,4,5-trifluorophenyl)bicyclohexyl

Precaución: for Research Use Only

Formula: C₂₀H₂₇F₃
MW: 324.43 g/mol
MDL Number: MFCD12911825
CAS Number: 139215-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR