Di(trimethylolpropane) 98% (by GC), (Chunks or flakes)

Supplier: Ambeed
Warning

Synonyms: 2,2'-Oxybis(methylene)bis(2-ethyl-1,3-propanediol), 2,2'-(Oxybis(methylene))bis(2-ethylpropane-1,3-diol), Bis[2-ethyl-2,2-bis(hydroxymethyl)ethyl] Ether

A1493480-100G A1493480-25G A1493480-500G A1493480-1000G
76665-954EA 12.74 USD
76665-954 76665-956 76665-958 77549-856
Di(trimethylolpropane) 98% (by GC), (Chunks or flakes)
Di(trimethylolpropane)

Precaución: for Research Use Only

Formula: C₁₂H₂₆O₅
MW: 250.34 g/mol
MDL Number: MFCD00192117
CAS Number: 23235-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR