N-Methyl-N-octanoyl-D-glucamine (MEGA-8) 97+%, (Chunks or flakes)

Supplier: Ambeed
Warning

A1493384-1G A1493384-250MG A1493384-5G
76665-948EA 44 USD
76665-948 76665-950 76665-952
N-Methyl-N-octanoyl-D-glucamine (MEGA-8) 97+%, (Chunks or flakes)
N-Methyl-N-octanoyl-D-glucamine (MEGA-8)

Precaución: for Research Use Only

Formula: C₁₅H₃₁NO₆
MDL Number: MFCD00134152
CAS Number: 85316-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR