3-(1-Azepanyl)-1-propanamine 93%

Supplier: Ambeed
Danger

Synonyms: 3-(Azepan-1-yl)propan-1-amine

A1493125-100MG A1493125-250MG A1493125-5G
76665-942EA 20.16 USD
76665-942 76665-944 76665-946
3-(1-Azepanyl)-1-propanamine 93%
3-(1-Azepanyl)-1-propanamine

Precaución: for Research Use Only

Formula: C₉H₂₀N₂
MW: 156.27 g/mol
MDL Number: MFCD00020990
CAS Number: 3437-33-0
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR