2-((Perfluoropropan-2-yl)oxy)isoindoline-1,3-dione 97%

Supplier: Ambeed
Warning

A1492939-100MG A1492939-1G A1492939-250MG A1492939-5G
76665-934EA 28.65 USD
76665-934 76665-936 76665-938 76665-940
2-((Perfluoropropan-2-yl)oxy)isoindoline-1,3-dione 97%
2-((Perfluoropropan-2-yl)oxy)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₁H₄F₇NO₃
MW: 331.14 g/mol
CAS Number: 2719854-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR