P(t-Bu)3 Pd G3 98%

Supplier: Ambeed
Warning

A1492810-100MG A1492810-1G A1492810-250MG A1492810-5G A1492810-25G
76665-926EA 10 USD
76665-926 76665-928 76665-930 77549-852 77875-479
P(t-Bu)3 Pd G3 98%
P(t-Bu)3 Pd G3

Precaución: for Research Use Only

Formula: C₂₅H₄₀NO₃PPdS
MDL Number: MFCD29037023
CAS Number: 1445086-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR